siRNA Details
| Gene Symbol: LOC100507437 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125384.1 | 3 | 1056 | |||
| NR_125385.1 | 2 | 921 | ||||
| GenBank mRNA | AK123017.1 | - | 2 | 921 | ||
| BU850282.1 | - | 3 | 584 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100507437| Gene Symbol: LOC100507437 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125384.1 | 3 | 1056 | |||
| NR_125385.1 | 2 | 921 | ||||
| GenBank mRNA | AK123017.1 | - | 2 | 921 | ||
| BU850282.1 | - | 3 | 584 | |||