siRNA Details
| Gene Symbol: RBM15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK091279.1 | - | 1 | 960 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK125665.1 | - | 2 | 1656 | Chr. 1 - 110368399 - 110373003 [+] on Build GRCh38 | |

RBM15
Genomic Map