siRNA Details
| Gene Symbol: TMEM39A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK092064.1 | - | 1 | 772 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BE939300.1 | - | 1 | 164 | Chr. 3 - 119429165 - 119429642 [+] on Build GRCh38 | |

TMEM39A
Genomic Map