siRNA Details
| Gene Symbol: HIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY358103.1 | - | 1 | 274 | ||
| JX965370.1 | - | 2 | 442 | |||
| JX965371.1 | - | 2 | 324 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIP1| Gene Symbol: HIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY358103.1 | - | 1 | 274 | ||
| JX965370.1 | - | 2 | 442 | |||
| JX965371.1 | - | 2 | 324 | |||