siRNA Details
| Gene Symbol: MIR3663HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121650.1 | 2 | 342 | |||
| NR_121651.1 | 2 | 342 | ||||
| GenBank mRNA | BC039338.1 | - | 2 | 342 | ||
| HY017741.1 | - | 2 | 335 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI463834.1 | - | 2 | 347 | Chr. 10 - 117160167 - 117169057 [-] on Build GRCh38 | |

MIR3663HG
Genomic Map