siRNA Details
| Gene Symbol: MIR3663HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121650.1 | 2 | 250 | |||
| NR_121651.1 | 2 | 250 | ||||
| GenBank mRNA | BC039338.1 | - | 2 | 250 | ||
| HY017741.1 | - | 2 | 243 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR3663HG| Gene Symbol: MIR3663HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121650.1 | 2 | 250 | |||
| NR_121651.1 | 2 | 250 | ||||
| GenBank mRNA | BC039338.1 | - | 2 | 250 | ||
| HY017741.1 | - | 2 | 243 | |||