siRNA Details
| Gene Symbol: INVS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC110864.1 | - | 2 | 663 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM804824.1 | - | 2 | 686 | Chr. 9 - 100099275 - 100304927 [+] on Build GRCh38 | |

INVS
Genomic Map