siRNA Details
| Gene Symbol: LINC01416 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110755.1 | 4 | 1398 | |||
| GenBank mRNA | BC043263.2 | - | 4 | 1398 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000563553.1 | - | 4 | 1398 | Chr. 18 - 55881688 - 55920199 [-] on Build GRCh38 | |

LINC01416
Genomic Map