siRNA Details
| Gene Symbol: LINC01416 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110755.1 | 4 | 357 | |||
| GenBank mRNA | BC043263.2 | - | 4 | 357 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI560997.1 | - | 4 | 362 | Chr. 18 - 55883171 - 55920201 [-] on Build GRCh38 | |

LINC01416
Genomic Map