siRNA Details
| Gene Symbol: DPCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK296220.1 | - | 4 | 1226 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK130871.1 | - | 3 | 454 | Chr. 10 - 101587215 - 101588879 [+] on Build GRCh38 | |

DPCD
Genomic Map