siRNA Details
| Gene Symbol: LOC401261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134654.1 | 2 | 2676 | |||
| GenBank mRNA | AK024196.1 | - | 2 | 1663 | ||
| AK098234.1 | - | 2 | 2675 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC401261| Gene Symbol: LOC401261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134654.1 | 2 | 2676 | |||
| GenBank mRNA | AK024196.1 | - | 2 | 1663 | ||
| AK098234.1 | - | 2 | 2675 | |||