siRNA Details
| Gene Symbol: LOC401261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK024196.1 | - | 2 | 711 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AU122601.2 | - | 2 | 711 | Chr. 6 - 42789260 - 42790339 [+] on Build GRCh38 | |

LOC401261
Genomic Map