siRNA Details
| Gene Symbol: TMTC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017020006.1 | XP_016875495.1 | 14 | 6787 | ||
| GenBank mRNA | AK097137.1 | - | 1 | 1811 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMTC1| Gene Symbol: TMTC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017020006.1 | XP_016875495.1 | 14 | 6787 | ||
| GenBank mRNA | AK097137.1 | - | 1 | 1811 | ||