siRNA Details
| Gene Symbol: CLSPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190481.1 | NP_001177410.1 | 24 | 5331 | ||
| NM_022111.3 | NP_071394.2 | 25 | 5523 | |||
| XM_011541938.2 | XP_011540240.1 | 25 | 5546 | |||
| XM_011541941.2 | XP_011540243.1 | 24 | 5378 | |||

CLSPN
Genomic Map