siRNA Details
| Gene Symbol: MTERF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017003380.1 | XP_016858869.1 | 8 | 3486 | ||
| XM_017003385.1 | XP_016858874.1 | 9 | 3322 | |||
| GenBank mRNA | AK126322.1 | - | 1 | 97 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MTERF4| Gene Symbol: MTERF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017003380.1 | XP_016858869.1 | 8 | 3486 | ||
| XM_017003385.1 | XP_016858874.1 | 9 | 3322 | |||
| GenBank mRNA | AK126322.1 | - | 1 | 97 | ||