siRNA Details
Gene Symbol: MTERF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003380.1 | XP_016858869.1 | 8 | 4401 | ||
XM_017003385.1 | XP_016858874.1 | 9 | 4237 | |||
GenBank mRNA | AK126322.1 | - | 1 | 1012 |
