siRNA Details
| Gene Symbol: SLC16A1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103743.1 | 2 | 1364 | |||
| NR_103744.1 | 2 | 598 | ||||
| GenBank mRNA | BC047723.1 | - | 2 | 584 | ||
| BQ708590.1 | - | 2 | 461 | |||
| BX648855.1 | - | 2 | 1318 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC16A1-AS1| Gene Symbol: SLC16A1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103743.1 | 2 | 1364 | |||
| NR_103744.1 | 2 | 598 | ||||
| GenBank mRNA | BC047723.1 | - | 2 | 584 | ||
| BQ708590.1 | - | 2 | 461 | |||
| BX648855.1 | - | 2 | 1318 | |||