siRNA Details
| Gene Symbol: LOC285766 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126020.1 | 2 | 836 | |||
| GenBank mRNA | AK097546.1 | - | 2 | 836 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC285766| Gene Symbol: LOC285766 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126020.1 | 2 | 836 | |||
| GenBank mRNA | AK097546.1 | - | 2 | 836 | ||