siRNA Details
| Gene Symbol: LINC01094 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC040182.2 | - | 3 | 2100 | ||
| BC044613.1 | - | 3 | 2379 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01094| Gene Symbol: LINC01094 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC040182.2 | - | 3 | 2100 | ||
| BC044613.1 | - | 3 | 2379 | |||