siRNA Details
Gene Symbol: LOC105370641 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011537447.1 | XP_011535749.1 | 1 | 1722 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK000041.1 | - | 11 | Chr. 14 - 95460372 - 95464213 [-] on Build GRCh38 |
