siRNA Details
| Gene Symbol: LOC105370641 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011537447.1 | XP_011535749.1 | 1 | 1809 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK000041.1 | - | 1 | 98 | Chr. 14 - 95460372 - 95464213 [-] on Build GRCh38 | |

LOC105370641
Genomic Map