siRNA Details
| Gene Symbol: PSPH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK054677.1 | - | 5 | 1498 | ||
| BC052958.1 | - | 3 | 2428 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ424231.1 | - | 1 | 427 | Chr. 7 - 56040373 - 56041201 [-] on Build GRCh38 | |

PSPH
Genomic Map