siRNA Details
| Gene Symbol: EBLN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036592.1 | 2 | 1585 | |||
| GenBank mRNA | AK091718.1 | - | 2 | 1585 | ||
| BC032471.1 | - | 2 | 1484 | |||
| BC033624.1 | - | 2 | 1484 | |||
| BC080614.1 | - | 2 | 1463 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EBLN3| Gene Symbol: EBLN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036592.1 | 2 | 1585 | |||
| GenBank mRNA | AK091718.1 | - | 2 | 1585 | ||
| BC032471.1 | - | 2 | 1484 | |||
| BC033624.1 | - | 2 | 1484 | |||
| BC080614.1 | - | 2 | 1463 | |||