siRNA Details
| Gene Symbol: LOC105374952 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134616.1 | 1 | 1092 | |||
| GenBank mRNA | BC035502.2 | - | 1 | 965 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105374952| Gene Symbol: LOC105374952 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134616.1 | 1 | 1092 | |||
| GenBank mRNA | BC035502.2 | - | 1 | 965 | ||