siRNA Details
| Gene Symbol: LOC105374952 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134616.1 | 1 | 562 | |||
| GenBank mRNA | BC035502.2 | - | 1 | 435 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ923201.1 | - | 1 | 505 | Chr. 6 - 17706189 - 17706996 [+] on Build GRCh38 | |

LOC105374952
Genomic Map