siRNA Details
| Gene Symbol: LINC00654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015406.2 | 2 | 3866 | |||
| GenBank mRNA | AL137476.1 | - | 1 | 3006 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000379053.4 | - | 3883 | Chr. 20 - 5500331 - 5504613 [-] on Build GRCh38 | ||

LINC00654
Genomic Map