siRNA Details
| Gene Symbol: HMGCS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098272.2 | NP_001091742.1 | 11 | 4693 | ||
| NM_001324219.1 | NP_001311148.1 | 10 | 4680 | |||
| NM_001324220.1 | NP_001311149.1 | 11 | 4713 | |||
| NM_001324222.1 | NP_001311151.1 | 10 | 4567 | |||
| NM_001324223.1 | NP_001311152.1 | 9 | 4508 | |||
| NM_001324224.1 | NP_001311153.1 | 10 | 4587 | |||
| NM_002130.7 | NP_002121.4 | 10 | 4634 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR605684.1 | - | 1 | 1128 | Chr. 5 - 43287699 - 43289354 [-] on Build GRCh38 | |

HMGCS1
Genomic Map