siRNA Details
| Gene Symbol: HMGCS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098272.2 | NP_001091742.1 | 11 | 4877 | ||
| NM_001324219.1 | NP_001311148.1 | 10 | 4864 | |||
| NM_001324220.1 | NP_001311149.1 | 11 | 4897 | |||
| NM_001324222.1 | NP_001311151.1 | 10 | 4751 | |||
| NM_001324223.1 | NP_001311152.1 | 9 | 4692 | |||
| NM_001324224.1 | NP_001311153.1 | 10 | 4771 | |||
| NM_002130.7 | NP_002121.4 | 10 | 4818 | |||

HMGCS1
Genomic Map