siRNA Details
Gene Symbol: HMGCS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098272.2 | NP_001091742.1 | 11 | 4084 | ||
NM_001324219.1 | NP_001311148.1 | 10 | 4071 | |||
NM_001324220.1 | NP_001311149.1 | 11 | 4104 | |||
NM_001324222.1 | NP_001311151.1 | 10 | 3958 | |||
NM_001324223.1 | NP_001311152.1 | 9 | 3899 | |||
NM_001324224.1 | NP_001311153.1 | 10 | 3978 | |||
NM_002130.7 | NP_002121.4 | 10 | 4025 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR605684.1 | - | 1 | 519 | Chr. 5 - 43287699 - 43289354 [-] on Build GRCh38 |
