siRNA Details
| Gene Symbol: LINC01618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040106.1 | 3 | 256 | |||
| GenBank mRNA | AL080225.1 | - | 3 | 256 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01618| Gene Symbol: LINC01618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040106.1 | 3 | 256 | |||
| GenBank mRNA | AL080225.1 | - | 3 | 256 | ||