siRNA Details
| Gene Symbol: LOC101928012 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110158.1 | 8 | 1008 | |||
| GenBank mRNA | BC041883.1 | - | 8 | 1011 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928012| Gene Symbol: LOC101928012 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110158.1 | 8 | 1008 | |||
| GenBank mRNA | BC041883.1 | - | 8 | 1011 | ||