siRNA Details
| Gene Symbol: LINC00403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK055145.1 | - | 1 | 635 | ||
| AK091058.1 | - | 5 | 1822 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00403| Gene Symbol: LINC00403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK055145.1 | - | 1 | 635 | ||
| AK091058.1 | - | 5 | 1822 | |||