siRNA Details
| Gene Symbol: LINC00403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK055145.1 | - | 1 | 987 | ||
| AK091058.1 | - | 5 | 2175 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AW293902.1 | - | 1 | 25 | Chr. 13 - 112109788 - 112110423 [+] on Build GRCh38 | |

LINC00403
Genomic Map