siRNA Details
| Gene Symbol: LOC101927460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109887.1 | 4 | 941 | |||
| GenBank mRNA | BC039471.1 | - | 4 | 940 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927460| Gene Symbol: LOC101927460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109887.1 | 4 | 941 | |||
| GenBank mRNA | BC039471.1 | - | 4 | 940 | ||