siRNA Details
| Gene Symbol: LOC101927460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109887.1 | 4 | 339 | |||
| GenBank mRNA | BC039471.1 | - | 4 | 339 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM473236.1 | - | 4 | 338 | Chr. 5 - 125493262 - 125601819 [+] on Build GRCh38 | |

LOC101927460
Genomic Map