siRNA Details
| Gene Symbol: LINC01343 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038928.1 | 3 | 425 | |||
| GenBank mRNA | BC042675.2 | - | 2 | 376 | ||
| BI827914.1 | - | 3 | 426 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01343| Gene Symbol: LINC01343 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038928.1 | 3 | 425 | |||
| GenBank mRNA | BC042675.2 | - | 2 | 376 | ||
| BI827914.1 | - | 3 | 426 | |||