siRNA Details
| Gene Symbol: LOC105377371 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC037784.2 | - | 1 | 989 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105377371| Gene Symbol: LOC105377371 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC037784.2 | - | 1 | 989 | ||