siRNA Details
| Gene Symbol: RCSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322923.1 | NP_001309852.1 | 6 | 2984 | ||
| NM_001322924.1 | NP_001309853.1 | 5 | 2912 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR597367.1 | - | 1 | 616 | Chr. 1 - 167705673 - 167707280 [+] on Build GRCh38 | |

RCSD1
Genomic Map