siRNA Details
| Gene Symbol: RPL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX537495.1 | - | 2 | 975 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPL15| Gene Symbol: RPL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX537495.1 | - | 2 | 975 | ||