siRNA Details
| Gene Symbol: KCNA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109845.1 | 3 | 5934 | |||
| NR_109846.1 | 3 | 6013 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNA3| Gene Symbol: KCNA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109845.1 | 3 | 5934 | |||
| NR_109846.1 | 3 | 6013 | ||||