siRNA Details
| Gene Symbol: KCNA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109845.1 | 3 | 6092 | |||
| NR_109846.1 | 3 | 6171 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR608017.1 | - | 1 | 1491 | Chr. 1 - 110653587 - 110655325 [-] on Build GRCh38 | |

KCNA3
Genomic Map