siRNA Details
| Gene Symbol: SIX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL540911.3 | - | 1 | 457 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR622406.1 | - | 1 | 457 | Chr. 19 - 45768387 - 45769685 [+] on Build GRCh38 | |

SIX5
Genomic Map