siRNA Details
| Gene Symbol: LOC101927078 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130785.1 | 10, 11 | 1491 | |||
| GenBank mRNA | AK097686.1 | - | 10, 11 | 1486 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB515172.1 | - | 2, 3 | 242 | Chr. 5 - 114447519 - 114472351 [-] on Build GRCh38 | |

LOC101927078
Genomic Map