siRNA Details
| Gene Symbol: LOC101927078 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130785.1 | 12 | 1636 | |||
| GenBank mRNA | AK097686.1 | - | 12 | 1631 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927078| Gene Symbol: LOC101927078 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130785.1 | 12 | 1636 | |||
| GenBank mRNA | AK097686.1 | - | 12 | 1631 | ||