siRNA Details
| Gene Symbol: CCDC148-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038850.1 | 5 | 556 | |||
| GenBank mRNA | BC021739.1 | - | 5 | 561 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG720090.1 | - | 5 | 561 | Chr. 2 - 158166650 - 158235978 [+] on Build GRCh38 | |

CCDC148-AS1
Genomic Map