siRNA Details
| Gene Symbol: CCDC148-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038850.1 | 5 | 763 | |||
| GenBank mRNA | BC021739.1 | - | 5 | 768 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC148-AS1| Gene Symbol: CCDC148-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038850.1 | 5 | 763 | |||
| GenBank mRNA | BC021739.1 | - | 5 | 768 | ||