siRNA Details
| Gene Symbol: C6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006714496.3 | XP_006714559.1 | 12 | 2013 | ||
| XM_011514114.2 | XP_011512416.1 | 12 | 2013 | |||
| XM_011514115.2 | XP_011512417.1 | 12 | 1934 | |||
| XM_011514116.2 | XP_011512418.1 | 13 | 2050 | |||
| XM_011514117.2 | XP_011512419.1 | 13 | 2092 | |||
| XM_011514118.2 | XP_011512420.1 | 12 | 1830 | |||
| XM_011514121.2 | XP_011512423.1 | 7 | 1099 | |||
| XM_017009818.1 | XP_016865307.1 | 13 | 2073 | |||
| XM_017009819.1 | XP_016865308.1 | 8 | 1276 | |||
| GenBank mRNA | J05063.1 | - | 1 | 702 | ||

C6
Genomic Map