siRNA Details
| Gene Symbol: LOC101929431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120660.1 | 3 | 3199 | |||
| GenBank mRNA | BX537934.1 | - | 3 | 3199 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929431| Gene Symbol: LOC101929431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120660.1 | 3 | 3199 | |||
| GenBank mRNA | BX537934.1 | - | 3 | 3199 | ||