siRNA Details
| Gene Symbol: LOC101926942 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110657.1 | 7 | 768 | |||
| GenBank mRNA | BC030278.1 | - | 7 | 768 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101926942| Gene Symbol: LOC101926942 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110657.1 | 7 | 768 | |||
| GenBank mRNA | BC030278.1 | - | 7 | 768 | ||