siRNA Details
| Gene Symbol: LOC101926942 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110657.1 | 3 | 279 | |||
| GenBank mRNA | BC030278.1 | - | 3 | 279 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI830277.1 | - | 3 | 279 | Chr. 10 - 90402644 - 90540805 [-] on Build GRCh38 | |

LOC101926942
Genomic Map